Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAAGTCTCTGCTTCTCAATGGC[C/T]TCCTCCAGCGTGAGGCCCACCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604853 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL28 - mitochondrial ribosomal protein L28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006428.4 | 663 | Silent Mutation | GAA,GAG | E,E 210 | NP_006419.2 | |
XM_005255041.1 | 663 | Silent Mutation | GAA,GAG | E,E 210 | XP_005255098.1 | |
XM_011522351.2 | 663 | Silent Mutation | GAA,GAG | E,E 210 | XP_011520653.1 |
TMEM8A - transmembrane protein 8A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |