Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCGGGCATTTTCTAAACAGGGT[C/T]GTTTCCCGCGTGCTCCTGCTTCGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610886 MIM: 601489 MIM: 610779 MIM: 611659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EME2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EME2 - essential meiotic structure-specific endonuclease subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGFALS - insulin like growth factor binding protein acid labile subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUBP2 - nucleotide binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284501.1 | Intron | NP_001271430.1 | ||||
NM_001284502.1 | Intron | NP_001271431.1 | ||||
NM_012225.3 | Intron | NP_036357.1 | ||||
XM_005255027.2 | Intron | XP_005255084.1 | ||||
XM_005255030.1 | Intron | XP_005255087.1 | ||||
XM_011522338.2 | Intron | XP_011520640.1 | ||||
XM_017022831.1 | Intron | XP_016878320.1 | ||||
XM_017022832.1 | Intron | XP_016878321.1 |
SPSB3 - splA/ryanodine receptor domain and SOCS box containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324081.1 | Intron | NP_001311010.1 | ||||
NM_080861.3 | Intron | NP_543137.2 | ||||
XM_005255673.4 | Intron | XP_005255730.1 |