Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAACAATTCCCAGGAGGGAGAC[A/G]CCAACTTCTCTTAGTGTGGAGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614604 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZDHHC7 - zinc finger DHHC-type containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145548.1 | 2169 | UTR 3 | NP_001139020.1 | |||
NM_017740.2 | 2169 | UTR 3 | NP_060210.2 | |||
XM_011523222.1 | 2169 | Intron | XP_011521524.1 | |||
XM_017023434.1 | 2169 | UTR 3 | XP_016878923.1 | |||
XM_017023435.1 | 2169 | UTR 3 | XP_016878924.1 | |||
XM_017023436.1 | 2169 | UTR 3 | XP_016878925.1 |