Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTAGGTCCATACCTCTGAATGAG[A/G]AGTCATCTGACAACAGGAGAATGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182888 | ||||||||||||||||||||
Literature Links: |
ZP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZP2 - zona pellucida glycoprotein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290104.1 | 2456 | Missense Mutation | NP_001277033.1 | |||
NM_003460.2 | 2456 | Missense Mutation | NP_003451.1 | |||
XM_011545942.2 | 2456 | Missense Mutation | XP_011544244.1 |