Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGGGCAAAGGGGAGGCTTCTCA[C/T]GAGCAGGGCTTCGGAGATGACCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX6A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX6A2 - cytochrome c oxidase subunit 6A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF843 - zinc finger protein 843 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136509.1 | 728 | Missense Mutation | ATG,GTG | M,V 1 | NP_001129981.1 |