Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGCCAGCATGTCGATGATGTG[A/G]GCCTCGAAGTCCGCGTCGTGCACGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607188 | ||||||||||||||||||||
Literature Links: |
DDX19A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX19A - DEAD-box helicase 19A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST3GAL2 - ST3 beta-galactoside alpha-2,3-sialyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006927.3 | 2139 | Silent Mutation | GCC,GCT | A,A 332 | NP_008858.1 |