Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGAACTGTAAGAGGTCCATGG[G/T]AGAGGCAGGGGAGGGAAGCACAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607886 MIM: 607887 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMTM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CMTM3 - CKLF like MARVEL transmembrane domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CMTM4 - CKLF like MARVEL transmembrane domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178818.2 | Intron | NP_848933.1 | ||||
NM_181521.2 | Intron | NP_852662.1 | ||||
XM_011522882.1 | Intron | XP_011521184.1 | ||||
XM_017022954.1 | Intron | XP_016878443.1 |