Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCATGGCAGTACCCAGCTTGCG[A/G]CAGTAGCAGAAGGCATTGAAGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602311 MIM: 607028 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGRP - agouti related neuropeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001138.1 | 657 | Silent Mutation | TGC,TGT | C,C 119 | NP_001129.1 |
ATP6V0D1 - ATPase H+ transporting V0 subunit d1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |