Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCCACCCCTCATCGCCCCTCCT[C/T]TGCGGTGGGCGGCTGCAGGTGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601615 MIM: 606447 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA3 - ATP binding cassette subfamily A member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001089.2 | 5813 | Missense Mutation | NP_001080.2 |
LOC106660606 - uncharacterized LOC106660606 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3677 - microRNA 3677 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4717 - microRNA 4717 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR940 - microRNA 940 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNPS1 - RNA binding protein with serine rich domain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |