Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGGTCTTTGCCAGGATGGTCTT[A/G]AAGATGTAGAATTTATCGGGATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604992 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRL - sarcalumenin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098814.1 | 1202 | Silent Mutation | TTC,TTT | F,F 381 | NP_001092284.1 | |
NM_001323667.1 | 1202 | Silent Mutation | TTC,TTT | F,F 339 | NP_001310596.1 | |
NM_001323668.1 | 1202 | Silent Mutation | TTC,TTT | F,F 339 | NP_001310597.1 | |
XM_017023527.1 | 1202 | Silent Mutation | TTC,TTT | F,F 765 | XP_016879016.1 | |
XM_017023528.1 | 1202 | Silent Mutation | TTC,TTT | F,F 339 | XP_016879017.1 | |
XM_017023529.1 | 1202 | Silent Mutation | TTC,TTT | F,F 339 | XP_016879018.1 |