Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCAATGACAAGGCCAGGGAGCC[A/G]AGCGTGGGTGGCAGGTGGCGAGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP8 - aquaporin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169.2 | 142 | Silent Mutation | CCA,CCG | P,P 20 | NP_001160.2 | |
XM_011545822.2 | 142 | Silent Mutation | CCA,CCG | P,P 21 | XP_011544124.1 | |
XM_011545823.2 | 142 | Silent Mutation | CCA,CCG | P,P 21 | XP_011544125.1 |