Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCAGAAAGCATATGTTCAGTTT[C/T]GTAACTGAAGGCTGTTGATAATTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616190 | ||||||||||||||||||||
Literature Links: |
CMTR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CMTR2 - cap methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099642.1 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | NP_001093112.1 | |
NM_001324374.1 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | NP_001311303.1 | |
NM_001324377.1 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | NP_001311306.1 | |
NM_001324378.1 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | NP_001311307.1 | |
NM_001324379.1 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | NP_001311308.1 | |
NM_018348.5 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | NP_060818.4 | |
XM_011523228.1 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | XP_011521530.1 | |
XM_011523229.1 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | XP_011521531.1 | |
XM_017023444.1 | 2856 | Missense Mutation | CAA,CGA | Q,R 769 | XP_016878933.1 |