Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGACAGCTTTTTGCAGTGGAC[C/T]CAGGCAATGATGTCACGGATATCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 140100 MIM: 140210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HP - haptoglobin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126102.2 | 122 | Missense Mutation | CCA,TCA | P,S 21 | NP_001119574.1 | |
NM_001318138.1 | 122 | Missense Mutation | CCA,TCA | P,S 21 | NP_001305067.1 | |
NM_005143.4 | 122 | Missense Mutation | CCA,TCA | P,S 21 | NP_005134.1 |
HPR - haptoglobin-related protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNL4B - thioredoxin like 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142317.1 | 122 | Intron | NP_001135789.1 | |||
NM_001142318.1 | 122 | Intron | NP_001135790.1 | |||
NM_001324354.1 | 122 | Intron | NP_001311283.1 | |||
NM_001324355.1 | 122 | Intron | NP_001311284.1 | |||
NM_017853.2 | 122 | Intron | NP_060323.1 | |||
XM_017023377.1 | 122 | Intron | XP_016878866.1 |