Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTTGCTTGGGTTGGTGGGCTC[C/G]GCCCGCACTGCCTGGTACTGCTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137070 MIM: 606838 | ||||||||||||||||||||
Literature Links: |
FUS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FUS - FUS RNA binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYCARD - PYD and CARD domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013258.4 | 482 | Silent Mutation | GCC,GCG | A,A 151 | NP_037390.2 | |
NM_145182.2 | 482 | Silent Mutation | GCC,GCG | A,A 132 | NP_660183.1 |
PYCARD-AS1 - PYCARD antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |