Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGTGAGGTCTCTTCCAGGACC[C/T]GTGCATCTCTTCCGACTCTCGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607838 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNPTG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNPTG - N-acetylglucosamine-1-phosphate transferase gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032520.4 | 266 | Silent Mutation | CCC,CCT | P,P 61 | NP_115909.1 | |
XM_017023782.1 | 266 | Silent Mutation | CCC,CCT | P,P 77 | XP_016879271.1 | |
XM_017023783.1 | 266 | UTR 5 | XP_016879272.1 |
TSR3 - TSR3, acp transferase ribosome maturation factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNKL - unkempt family like zinc finger | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |