Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAGCGCCTGACTCGAGAGCACC[A/G]CCTTTCAGTCCACTGGGACCAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 115442 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK2A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSNK2A2 - casein kinase 2 alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001896.2 | Intron | NP_001887.1 | ||||
XM_005255800.3 | Intron | XP_005255857.1 | ||||
XM_005255801.3 | Intron | XP_005255858.1 | ||||
XM_017022945.1 | Intron | XP_016878434.1 |