Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTGATCACAGATGAATGTAGGA[C/T]TGATGCAAGTCACTTCCAGGAACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604230 MIM: 601421 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAT1 - adenosine deaminase, tRNA specific 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KARS - lysyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130089.1 | 1513 | Missense Mutation | AAT,AGT | N,S 486 | NP_001123561.1 | |
NM_005548.2 | 1513 | Intron | NP_005539.1 | |||
XM_017023217.1 | 1513 | Missense Mutation | AAT,AGT | N,S 302 | XP_016878706.1 |