Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAATGACACTGTAACTGCATAAA[A/G]CCAAAGAAAGCTAGAAAAAGGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610937 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPGRIP1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPGRIP1L - RPGRIP1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005255867.1 | 5804 | UTR 3 | XP_005255924.1 | |||
XM_005255868.1 | 5804 | UTR 3 | XP_005255925.1 | |||
XM_011522970.1 | 5804 | Intron | XP_011521272.1 | |||
XM_011522971.2 | 5804 | Intron | XP_011521273.1 | |||
XM_011522973.2 | 5804 | Intron | XP_011521275.1 | |||
XM_017023094.1 | 5804 | UTR 3 | XP_016878583.1 | |||
XM_017023095.1 | 5804 | UTR 3 | XP_016878584.1 | |||
XM_017023096.1 | 5804 | Intron | XP_016878585.1 | |||
XM_017023097.1 | 5804 | Intron | XP_016878586.1 | |||
XM_017023098.1 | 5804 | UTR 3 | XP_016878587.1 | |||
XM_017023099.1 | 5804 | UTR 3 | XP_016878588.1 | |||
XM_017023100.1 | 5804 | Intron | XP_016878589.1 |