Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCCATGGACTTTGAAAACCAGC[A/G]CACACTTAGGTTTGAATTCTTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2I PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2I - ubiquitin conjugating enzyme E2 I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003345.4 | Intron | NP_003336.1 | ||||
NM_194259.2 | Intron | NP_919235.1 | ||||
NM_194260.2 | Intron | NP_919236.1 | ||||
NM_194261.2 | Intron | NP_919237.1 | ||||
XM_005255540.2 | Intron | XP_005255597.1 | ||||
XM_005255541.4 | Intron | XP_005255598.1 | ||||
XM_011522644.2 | Intron | XP_011520946.1 | ||||
XM_011522645.1 | Intron | XP_011520947.1 | ||||
XM_017023640.1 | Intron | XP_016879129.1 |