Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTGTGTCGATGATGGGCACAGC[A/G]AGTTTCTGCAGGATCCGCGGTTCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611285 MIM: 608018 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD5 - potassium channel tetramerization domain containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRSS27 - protease, serine 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318395.1 | 663 | Silent Mutation | CTC,CTT | L,L 79 | NP_001305324.1 | |
NM_031948.4 | 663 | Silent Mutation | CTC,CTT | L,L 181 | NP_114154.1 | |
XM_011522689.2 | 663 | Silent Mutation | CTC,CTT | L,L 79 | XP_011520991.1 |