Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GTTATAGCGTCCAGGTCCAACACAG[C/G]ATATTTGCATACAAAGCCACTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607974 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NETO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NETO2 - neuropilin and tolloid like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201477.1 | 3228 | UTR 3 | NP_001188406.1 | |||
NM_018092.4 | 3228 | UTR 3 | NP_060562.3 | |||
XM_006721289.1 | 3228 | UTR 3 | XP_006721352.1 | |||
XM_006721291.1 | 3228 | UTR 3 | XP_006721354.1 | |||
XM_006721292.1 | 3228 | Intron | XP_006721355.1 | |||
XM_017023740.1 | 3228 | UTR 3 | XP_016879229.1 |