Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTGCAGGATAAGGCTTCCTTA[C/T]GTAGTGAACGGTTGAGAATATTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSDL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSDL1 - hydroxysteroid dehydrogenase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146051.1 | 984 | Missense Mutation | NP_001139523.1 | |||
NM_031463.4 | 984 | Missense Mutation | NP_113651.4 | |||
XM_005256189.2 | 984 | Missense Mutation | XP_005256246.1 |
LOC729887 - uncharacterized LOC729887 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MBTPS1 - membrane bound transcription factor peptidase, site 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |