Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCTACAAGTCTCCCGAGGTCTC[C/T]GGCGTTTCCTTCTCCAGATGCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606216 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSRB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSRB1 - methionine sulfoxide reductase B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL3L - ribosomal protein L3 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005061.2 | 1710 | Silent Mutation | CCA,CCG | P,P 401 | NP_005052.1 | |
XM_011522571.2 | 1710 | Silent Mutation | CCA,CCG | P,P 406 | XP_011520873.1 |