Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAAATGTCCATCATAACAAATA[C/T]AAAATGTTTCCAGAAAGTATCATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605943 | ||||||||||||||||||||
Literature Links: |
GDE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDE1 - glycerophosphodiester phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324066.1 | 909 | Missense Mutation | ATA,GTA | I,V 139 | NP_001310995.1 | |
NM_001324067.1 | 909 | Missense Mutation | ATA,GTA | I,V 218 | NP_001310996.1 | |
NM_016641.3 | 909 | Missense Mutation | ATA,GTA | I,V 249 | NP_057725.1 |
TMC5 - transmembrane channel like 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |