Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGCTGAGATCCTCTGTGCCTCT[A/G]GCCAGTCTTCACAGTAGTCACTACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604886 | ||||||||||||||||||||
Literature Links: |
EMC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EMC8 - ER membrane protein complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142288.1 | 432 | UTR 3 | NP_001135760.1 | |||
NM_006067.4 | 432 | Missense Mutation | CCA,CTA | P,L 165 | NP_006058.1 | |
XM_011522813.2 | 432 | Intron | XP_011521115.1 | |||
XM_017022867.1 | 432 | Missense Mutation | CCA,CTA | P,L 113 | XP_016878356.1 |