Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614666 MIM: 610998 MIM: 611118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC78 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM173A - family with sequence similarity 173 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271285.1 | 1593 | Intron | NP_001258214.1 | |||
NM_023933.2 | 1593 | Intron | NP_076422.1 |
HAGHL - hydroxyacylglutathione hydrolase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METRN - meteorin, glial cell differentiation regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NARFL - nuclear prelamin A recognition factor like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |