Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTGGTAGGCCTGGAAGACGCT[C/G]AGCAGATCCTTCATACCAGCCGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612960 MIM: 602698 | ||||||||||||||||||||
Literature Links: |
ESRP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESRP2 - epithelial splicing regulatory protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024939.2 | 2131 | Silent Mutation | CTC,CTG | L,L 681 | NP_079215.2 | |
XM_005256153.4 | 2131 | Silent Mutation | CTC,CTG | L,L 680 | XP_005256210.1 | |
XM_006721273.3 | 2131 | Silent Mutation | CTC,CTG | L,L 555 | XP_006721336.1 | |
XM_011523338.2 | 2131 | Silent Mutation | CTC,CTG | L,L 555 | XP_011521640.1 |
MIR6773 - microRNA 6773 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFATC3 - nuclear factor of activated T-cells 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |