Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCGCATGGTGACCGTCCCCCCA[C/T]GTCACTACTGCACAGTGGCCAACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605088 MIM: 612033 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MVP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MVP - major vault protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293204.1 | 359 | Missense Mutation | CGT,TGT | R,C 56 | NP_001280133.1 | |
NM_001293205.1 | 359 | Missense Mutation | CGT,TGT | R,C 56 | NP_001280134.1 | |
NM_005115.4 | 359 | Missense Mutation | CGT,TGT | R,C 56 | NP_005106.2 | |
NM_017458.3 | 359 | Missense Mutation | CGT,TGT | R,C 56 | NP_059447.2 |
PAGR1 - PAXIP1 associated glutamate rich protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |