Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTGGAGGAGAGAGAGGAAAGT[G/T]AAGGAAGAAAGAGGACTGTGGAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CES5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CES5A - carboxylesterase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143685.1 | 1829 | Missense Mutation | TTA,TTC | L,F 560 | NP_001137157.1 | |
NM_001190158.1 | 1829 | Missense Mutation | TTA,TTC | L,F 589 | NP_001177087.1 | |
NM_145024.2 | 1829 | Missense Mutation | TTA,TTC | L,F 510 | NP_659461.1 |