Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGATACTGCTCCAGTTCCAAGA[C/T]GAGGCTTCAAGCCTGCCCCTGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611416 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOX3 - TOX high mobility group box family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080430.3 | Intron | NP_001073899.2 | ||||
NM_001146188.2 | Intron | NP_001139660.1 | ||||
XM_005255892.3 | Intron | XP_005255949.1 | ||||
XM_011523002.2 | Intron | XP_011521304.1 | ||||
XM_017023142.1 | Intron | XP_016878631.1 |