Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCCACCCAGCCCGGGCAGGGCT[A/C]TTCCCAGCAGAGCAGTCAGCCCTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137070 | ||||||||||||||||||||
Literature Links: |
FUS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FUS - FUS RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170634.1 | 845 | Missense Mutation | TAT,TCT | Y,S 25 | NP_001164105.1 | |
NM_001170937.1 | 845 | Missense Mutation | TAT,TCT | Y,S 25 | NP_001164408.1 | |
NM_004960.3 | 845 | Missense Mutation | TAT,TCT | Y,S 25 | NP_004951.1 | |
XM_005255233.4 | 845 | UTR 5 | XP_005255290.1 | |||
XM_011545781.1 | 845 | Missense Mutation | TAT,TCT | Y,S 25 | XP_011544083.1 | |
XM_011545782.2 | 845 | Intron | XP_011544084.1 |