Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTGTAGCAAGTGTGCCCAGGG[C/G]TGTGTTTGCAAAGGGGCGTCAGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 156349 MIM: 156352 MIM: 156353 | ||||||||||||||||||||
Literature Links: |
MT1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MT1B - metallothionein 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MT1F - metallothionein 1F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301272.1 | 257 | UTR 3 | NP_001288201.1 | |||
NM_005949.3 | 257 | Silent Mutation | GGC,GGG | G,G 47 | NP_005940.1 |
MT1G - metallothionein 1G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |