Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACAGACTGCACTCCTGGTTGTC[C/T]TCGTCCTCCTTGCTGTGGCGCTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602957 | ||||||||||||||||||||
Literature Links: |
CCL22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCL22 - C-C motif chemokine ligand 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002990.4 | 88 | Missense Mutation | CTC,TTC | L,F 12 | NP_002981.2 | |
XM_017023531.1 | 88 | Missense Mutation | CTC,TTC | L,F 12 | XP_016879020.1 |