Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGCTGTCGGAGTGAATGCGCCG[G/T]TGACTGAGGAGGAGGGAAGAATAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF785 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF785 - zinc finger protein 785 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152458.6 | 1456 | Missense Mutation | CAA,CAC | Q,H 330 | NP_689671.2 | |
XM_011545752.2 | 1456 | Missense Mutation | CAA,CAC | Q,H 343 | XP_011544054.1 | |
XM_011545753.2 | 1456 | Missense Mutation | CAA,CAC | Q,H 343 | XP_011544055.1 | |
XM_017022967.1 | 1456 | Missense Mutation | CAA,CAC | Q,H 330 | XP_016878456.1 |