Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTGAAGTGGTCTCGGGAGAATC[A/G]GATCAGAGCATCACTGAAGCTGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600659 MIM: 614117 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
E2F4 - E2F transcription factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EXOC3L1 - exocyst complex component 3 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178516.3 | 1646 | Nonsense Mutation | CGA,TGA | R,* 469 | NP_848611.2 |
KIAA0895L - KIAA0895 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |