Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACCTGTTCATAGTCGTCAGCCC[A/G]AGCCAGCTGCGCCAGGCCCTCAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607028 MIM: 614232 | ||||||||||||||||||||
Literature Links: |
ATP6V0D1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP6V0D1 - ATPase H+ transporting V0 subunit d1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004691.4 | 761 | Missense Mutation | CGG,TGG | R,W 257 | NP_004682.2 | |
XM_011523444.1 | 761 | Missense Mutation | CGG,TGG | R,W 180 | XP_011521746.1 | |
XM_011523445.1 | 761 | Missense Mutation | CGG,TGG | R,W 180 | XP_011521747.1 | |
XM_011523446.1 | 761 | Missense Mutation | CGG,TGG | R,W 180 | XP_011521748.1 |
HSD11B2 - hydroxysteroid 11-beta dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |