Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGAGCTGCTGGAACTTGGACAA[C/T]AGGCACTCATCACCGCAGATCCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf95 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf95 - chromosome 16 open reading frame 95 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195124.1 | 574 | Silent Mutation | CTA,CTG | L,L 192 | NP_001182053.1 | |
NM_001195125.1 | 574 | Missense Mutation | TAT,TGT | Y,C 131 | NP_001182054.1 | |
NM_001256917.1 | 574 | Missense Mutation | TAT,TGT | Y,C 116 | NP_001243846.1 |