Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGACCCTCCACCGGGCTCGCCG[G/T]TCCGTCGGACTTTTGGCTGAGATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104155 | ||||||||||||||||||||
Literature Links: |
ZFHX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFHX3 - zinc finger homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164766.1 | 8410 | Missense Mutation | ACG,CCG | T,P 2761 | NP_001158238.1 | |
NM_006885.3 | 8410 | Missense Mutation | ACG,CCG | T,P 3675 | NP_008816.3 | |
XM_005255957.3 | 8410 | Missense Mutation | ACG,CCG | T,P 3674 | XP_005256014.1 | |
XM_017023251.1 | 8410 | Missense Mutation | ACG,CCG | T,P 2792 | XP_016878740.1 |