Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATGCCTTCTAAGCTGGGGGAG[A/G]CGGTACCTTCAGGGGACACTCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603436 | ||||||||||||||||||||
Literature Links: |
ZNF205 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF205 - zinc finger protein 205 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042428.1 | 279 | Missense Mutation | ACG,GCG | T,A 37 | NP_001035893.1 | |
NM_001278158.1 | 279 | Missense Mutation | ACG,GCG | T,A 37 | NP_001265087.1 | |
NM_003456.2 | 279 | Missense Mutation | ACG,GCG | T,A 37 | NP_003447.2 | |
XM_005255558.2 | 279 | Missense Mutation | ACG,GCG | T,A 37 | XP_005255615.1 |
ZNF205-AS1 - ZNF205 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |