Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGCGCTCCCGAACAGACCTGAC[A/G]TAGATCCGAAGTGGCCCGCGCCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608954 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MON1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MON1B - MON1 homolog B, secretory trafficking associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286639.1 | 116 | UTR 5 | NP_001273568.1 | |||
NM_001286640.1 | 116 | UTR 5 | NP_001273569.1 | |||
NM_014940.3 | 116 | UTR 5 | NP_055755.1 |
SYCE1L - synaptonemal complex central element protein 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |