Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAATATAAGTGTGATGCCGTAA[C/T]ATTTTCTTACATGTCAGAATACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600023 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH11 - cadherin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308392.1 | 3668 | UTR 3 | NP_001295321.1 | |||
NM_001797.3 | 3668 | UTR 3 | NP_001788.2 | |||
XM_005255761.3 | 3668 | UTR 3 | XP_005255818.1 | |||
XM_005255762.2 | 3668 | UTR 3 | XP_005255819.1 | |||
XM_005255763.2 | 3668 | UTR 3 | XP_005255820.1 | |||
XM_011522803.2 | 3668 | UTR 3 | XP_011521105.1 | |||
XM_017022830.1 | 3668 | UTR 3 | XP_016878319.1 |