Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGCTGTTCCGGTGGAATGTTTC[A/G]GACCTAGGTGGCCTGGGCTGTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107265 MIM: 611869 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD19 - CD19 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178098.1 | 244 | Silent Mutation | TCA,TCG | S,S 127 | NP_001171569.1 | |
NM_001770.5 | 244 | Silent Mutation | TCA,TCG | S,S 127 | NP_001761.3 | |
XM_006721103.3 | 244 | Silent Mutation | TCA,TCG | S,S 38 | XP_006721166.1 | |
XM_011545981.1 | 244 | Silent Mutation | TCA,TCG | S,S 127 | XP_011544283.1 | |
XM_017023893.1 | 244 | Silent Mutation | TCA,TCG | S,S 38 | XP_016879382.1 |
RABEP2 - rabaptin, RAB GTPase binding effector protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |