Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTGCTCCCGGATGGCCTCCTG[A/T]TCCTCTTGCTGCTGCTAATGCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612960 MIM: 609362 | ||||||||||||||||||||
Literature Links: |
ESRP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESRP2 - epithelial splicing regulatory protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G15 - phospholipase A2 group XV | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012320.3 | 111 | Missense Mutation | ATC,TTC | I,F 19 | NP_036452.1 | |
XM_005255886.3 | 111 | Missense Mutation | ATC,TTC | I,F 19 | XP_005255943.1 | |
XM_011522979.2 | 111 | Missense Mutation | ATC,TTC | I,F 19 | XP_011521281.1 | |
XM_011522980.2 | 111 | UTR 5 | XP_011521282.2 |