Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATCGGACTCCCCAAACCGGTCT[A/G]GGTGGCACTTGAGGCTCCCAGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603334 MIM: 611304 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAH3 - dynein axonemal heavy chain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017539.2 | Intron | NP_060009.1 | ||||
XM_006721062.1 | Intron | XP_006721125.1 | ||||
XM_011545882.2 | Intron | XP_011544184.1 | ||||
XM_011545883.1 | Intron | XP_011544185.1 | ||||
XM_011545885.2 | Intron | XP_011544187.1 | ||||
XM_011545886.2 | Intron | XP_011544188.1 | ||||
XM_011545888.2 | Intron | XP_011544190.1 | ||||
XM_011545889.2 | Intron | XP_011544191.1 | ||||
XM_017023426.1 | Intron | XP_016878915.1 | ||||
XM_017023427.1 | Intron | XP_016878916.1 | ||||
XM_017023428.1 | Intron | XP_016878917.1 | ||||
XM_017023429.1 | Intron | XP_016878918.1 | ||||
XM_017023430.1 | Intron | XP_016878919.1 | ||||
XM_017023431.1 | Intron | XP_016878920.1 | ||||
XM_017023432.1 | Intron | XP_016878921.1 |
TMEM159 - transmembrane protein 159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301769.1 | Intron | NP_001288698.1 | ||||
NM_001301771.1 | Intron | NP_001288700.1 | ||||
NM_001301773.1 | Intron | NP_001288702.1 | ||||
NM_001301774.1 | Intron | NP_001288703.1 | ||||
NM_001301775.1 | Intron | NP_001288704.1 | ||||
NM_020422.5 | Intron | NP_065155.3 | ||||
XM_005255439.4 | Intron | XP_005255496.1 | ||||
XM_006721066.2 | Intron | XP_006721129.1 | ||||
XM_006721067.3 | Intron | XP_006721130.1 | ||||
XM_006721069.3 | Intron | XP_006721132.1 | ||||
XM_017023471.1 | Intron | XP_016878960.1 | ||||
XM_017023472.1 | Intron | XP_016878961.1 | ||||
XM_017023473.1 | Intron | XP_016878962.1 | ||||
XM_017023474.1 | Intron | XP_016878963.1 |