Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGCCCGCTTTTCCATGGAGCGG[G/T]CAGAATTGGGACAGGCCTCCTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603243 MIM: 139311 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMFR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMFR - autocrine motility factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144.5 | 1836 | UTR 3 | NP_001135.3 | |||
NM_001323511.1 | 1836 | UTR 3 | NP_001310440.1 | |||
NM_001323512.1 | 1836 | UTR 3 | NP_001310441.1 | |||
XM_005255890.3 | 1836 | UTR 3 | XP_005255947.1 |
GNAO1 - G protein subunit alpha o1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |