Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTTCTCCATTTCCAGCAGGACC[C/T]GCTTGTGCTGCTCTGCGATGGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608333 MIM: 180663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOK4 - docking protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018110.3 | 999 | Missense Mutation | CAG,CGG | Q,R 236 | NP_060580.2 | |
XM_005256042.3 | 999 | Missense Mutation | CAG,CGG | Q,R 236 | XP_005256099.1 | |
XM_005256043.2 | 999 | Missense Mutation | CAG,CGG | Q,R 236 | XP_005256100.1 |
POLR2C - RNA polymerase II subunit C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |