Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGTTCTGAATGACGGAGATCTC[A/G]TTGGCCTTCATCGCCAGCCCGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602218 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371253 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371253 - uncharacterized LOC105371253 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SALL1 - spalt like transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127892.1 | 3781 | Silent Mutation | NP_001121364.1 | |||
NM_002968.2 | 3781 | Silent Mutation | NP_002959.2 | |||
XM_006721241.3 | 3781 | Silent Mutation | XP_006721304.1 | |||
XM_011523254.2 | 3781 | Silent Mutation | XP_011521556.1 |