Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGGGCTCCCAAAGAGCGCCCTG[C/T]GGAGCCTCTCACCCCTCCCCCATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615549 MIM: 182381 MIM: 602353 | ||||||||||||||||||||
Literature Links: |
ARMC5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMC5 - armadillo repeat containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC5A2 - solute carrier family 5 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGFB1I1 - transforming growth factor beta 1 induced transcript 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042454.2 | 89 | Missense Mutation | GCG,GTG | A,V 29 | NP_001035919.1 | |
NM_001164719.1 | 89 | Missense Mutation | GCG,GTG | A,V 12 | NP_001158191.1 | |
NM_015927.4 | 89 | Missense Mutation | GCG,GTG | A,V 12 | NP_057011.2 |