Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATCTTCAAGCCATTCTTACTGTA[C/T]GCTGTGATGGAGGGGATGCCTGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1G1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1G1 - adaptor related protein complex 1 gamma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030007.1 | 2441 | Silent Mutation | GCA,GCG | A,A 712 | NP_001025178.1 | |
NM_001128.5 | 2441 | Silent Mutation | GCA,GCG | A,A 709 | NP_001119.3 |