Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAACCCAGAATTTGCTAAGAGA[C/T]GGAGGAACCTCAGAAAAGCTATGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BFAR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BFAR - bifunctional apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016561.2 | 263 | Missense Mutation | ACG,ATG | T,M 1 | NP_057645.1 | |
XM_005255350.2 | 263 | UTR 5 | XP_005255407.1 | |||
XM_011522520.1 | 263 | Missense Mutation | ACG,ATG | T,M 1 | XP_011520822.1 |